Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PLAUR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005376.2 | NP_001005376.1 | 6 | 925 | ||
NM_001005377.2 | NP_001005377.1 | 5 | 790 | |||
NM_002659.3 | NP_002650.1 | 6 | 925 | |||
XM_005258990.4 | XP_005259047.1 | 6 | 747 | |||
XM_011527027.2 | XP_011525329.1 | 5 | 603 | |||
XM_011527028.2 | XP_011525330.1 | 6 | 767 | |||
XM_011527029.1 | XP_011525331.1 | 6 | 767 | |||
XM_017026872.1 | XP_016882361.1 | 6 | 1160 | |||
XM_017026873.1 | XP_016882362.1 | 4 | 813 | |||
GenBank mRNA | A18757.1 | - | 6 | 745 | ||
AK290774.1 | - | 6 | 745 | |||
AY029180.1 | - | 6 | 745 | |||
BC002788.2 | - | 6 | 764 | 3617894 | ||
CR456952.1 | - | 6 | 699 | |||
M83246.1 | - | 6 | 716 | |||
U08839.1 | - | 5 | 990 | |||
X51675.1 | - | 6 | 745 | |||
X74039.1 | - | 6 | 745 |