Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321386.1 | NP_001308315.1 | 5 | 1158 | ||
NM_001321387.1 | NP_001308316.1 | 5 | 1457 | |||
NM_002825.5 | NP_002816.1 | 5 | 1395 | |||
XM_017012473.1 | XP_016867962.1 | 5 | 1275 |