Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ZNF43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256648.1 | NP_001243577.1 | 7 | 3056 | ||
NM_001256649.1 | NP_001243578.1 | 7 | 2921 | |||
NM_001256650.1 | NP_001243579.1 | 6 | 2793 | |||
NM_001256651.1 | NP_001243580.1 | 3 | 2407 | |||
NM_001256653.1 | NP_001243582.1 | 4 | 2534 | |||
NM_001256654.1 | NP_001243583.1 | 3 | 2416 | |||
NM_003423.3 | NP_003414.2 | 4 | 2543 | |||
XM_011528257.2 | XP_011526559.1 | 4 | 2465 | |||
XM_011528259.2 | XP_011526561.1 | 4 | 3022 | |||
XM_017027207.1 | XP_016882696.1 | 7 | 8534 | |||
XM_017027208.1 | XP_016882697.1 | 7 | 3214 | |||
XM_017027209.1 | XP_016882698.1 | 7 | 8267 | |||
XM_017027210.1 | XP_016882699.1 | 6 | 8058 | |||
XM_017027211.1 | XP_016882700.1 | 7 | 8201 | |||
XM_017027212.1 | XP_016882701.1 | 6 | 3061 | |||
XM_017027213.1 | XP_016882702.1 | 7 | 8309 | |||
XM_017027214.1 | XP_016882703.1 | 5 | 2984 | |||
XM_017027215.1 | XP_016882704.1 | 6 | 2823 | |||
XM_017027216.1 | XP_016882705.1 | 5 | 2609 | |||
GenBank mRNA | AK291353.1 | - | 7 | 2871 | ||
BC006528.2 | - | 4 | 2510 | 3687640 | ||
BT009756.1 | - | 3 | 2355 | |||
CR933640.1 | - | 7 | 3016 | |||
X59244.1 | - | 6 | 2692 |