Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCL27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006664.3 | NP_006655.1 | 2 | 202 | ||
GenBank mRNA | AB010445.1 | - | 2 | 202 | ||
AF082393.1 | - | 2 | 143 | |||
AJ243542.1 | - | 2 | 167 | |||
BC148263.1 | - | 2 | 165 | 8860203 | ||
BM719357.1 | - | 1 | 75 |