Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BTN3A2 SIRNA Assays ›
Gene Symbol:
BTN3A2
Gene Name:
butyrophilin subfamily 3 member A2
Gene Aliases:
BT3.2, BTF4, BTN3.2, CD277
Chromosome Location:
Chr.6: 26365159 - 26451697 on Build GRCh38
UniGene ID:
Hs.376046
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
289635
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BTN3A2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001197246.2
NP_001184175.1
9
2447
NM_001197247.2
NP_001184176.1
11
3726
NM_001197248.2
NP_001184177.1
9
3283
NM_001197249.2
NP_001184178.1
10
3344
NM_007047.4
NP_008978.2
11
3434
XM_005248827.3
XP_005248884.1
11
3704
XM_005248831.3
XP_005248888.1
10
3221
XM_005248832.3
XP_005248889.1
10
3503
XM_006714979.3
XP_006715042.1
11
3437
XM_006714980.3
XP_006715043.1
11
3719
XM_006714981.3
XP_006715044.1
11
3427
XM_006714982.3
XP_006715045.1
10
3347
XM_011514267.2
XP_011512569.1
11
3303
XM_011514269.2
XP_011512571.1
10
3206
XM_011514270.2
XP_011512572.1
10
3140
XM_011514271.2
XP_011512573.1
10
3006
XM_017010211.1
XP_016865700.1
11
3412
XM_017010212.1
XP_016865701.1
11
3288
XM_017010213.1
XP_016865702.1
10
3309
XM_017010214.1
XP_016865703.1
10
3614
XM_017010215.1
XP_016865704.1
10
3422
XM_017010216.1
XP_016865705.1
9
3131
XM_017010217.1
XP_016865706.1
9
3050
GenBank mRNA
BC067086.1
-
1
1488
U90143.1
-
1
1390
Back To Top
More Information
Panther Classification:
Molecular Function -
immunoglobulin receptor superfamily
Gene Ontology Categories:
Function(s)
T cell mediated immunity
interferon-gamma secretion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-9ljwx:80/10.239.166.46:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0