Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PREX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020820.3 | NP_065871.2 | 40 | 5795 | ||
XM_011528934.2 | XP_011527236.1 | 32 | 4697 | |||
GenBank mRNA | AB037836.2 | - | 40 | 5658 | ||
AJ320261.1 | - | 40 | 5795 | |||
AK090860.1 | - | 1 | 1814 | |||
AL136579.1 | - | 23 | 3752 | |||
AL832913.1 | - | 33 | 4764 | |||
BC053616.1 | - | 36 | 5245 |