Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OBP2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293189.1 | NP_001280118.1 | 5 | 510 | ||
NM_001293193.1 | NP_001280122.1 | 4 | 376 | |||
NM_014582.2 | NP_055397.1 | 5 | 445 | |||
NR_120603.1 | 5 | 494 | ||||
XM_017014656.1 | XP_016870145.1 | 6 | 1386 | |||
GenBank mRNA | AJ251021.1 | - | 5 | 445 | ||
AJ251022.1 | - | 5 | 494 | |||
AJ251023.1 | - | 5 | 376 | |||
AJ251024.1 | - | 5 | 510 | |||
AM087898.1 | - | 5 | 361 | |||
BC069563.1 | - | 5 | 430 | 7262146 |