Search Thermo Fisher Scientific
Gene Symbol: ZNF687 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304763.1 | NP_001291692.1 | 10 | 5292 | ||
NM_001304764.1 | NP_001291693.1 | 9 | 4493 | |||
NM_020832.2 | NP_065883.1 | 9 | 4423 | |||
XM_005245366.3 | XP_005245423.1 | 9 | 4861 | |||
XM_011509811.2 | XP_011508113.1 | 8 | 4293 | |||
XM_011509812.2 | XP_011508114.1 | 10 | 4740 | |||
XM_011509813.2 | XP_011508115.1 | 9 | 4624 | |||
GenBank mRNA | AB037862.1 | - | 10 | 5292 | ||
AK023105.1 | - | 8 | 3191 | |||
BC032463.1 | - | 9 | 4420 | 5196247 | ||
CR749307.1 | - | 9 | 4494 |