Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSBPL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003712.1 | NP_001003712.1 | 15 | 2129 | ||
NM_001319652.1 | NP_001306581.1 | 18 | 2254 | |||
NM_001319653.1 | NP_001306582.1 | 16 | 2166 | |||
NM_001319655.1 | NP_001306584.1 | 13 | 2060 | |||
NM_020841.4 | NP_065892.1 | 16 | 2175 | |||
XM_005268621.4 | XP_005268678.1 | 19 | 2273 | |||
XM_006719224.2 | XP_006719287.1 | 15 | 2134 | |||
XM_011537855.2 | XP_011536157.1 | 16 | 1792 | |||
XM_011537856.2 | XP_011536158.1 | 17 | 2210 | |||
XM_011537857.2 | XP_011536159.1 | 19 | 2264 | |||
XM_017018768.1 | XP_016874257.1 | 16 | 1807 | |||
XM_017018769.1 | XP_016874258.1 | 17 | 2178 | |||
GenBank mRNA | AB040884.3 | - | 16 | 1846 | ||
AF323730.1 | - | 15 | 2129 | |||
AF392452.1 | - | 16 | 2176 | |||
AK092160.1 | - | 13 | 2061 | |||
AK289997.1 | - | 16 | 1895 | |||
AL832876.1 | - | 3 | 231 | |||
BC012935.1 | - | 2 | 160 | |||
BC082235.1 | - | 16 | 2114 | |||
BC093834.1 | - | 15 | 1774 | 7939679 | ||
BC101529.1 | - | 15 | 1774 | 8069035 | ||
BC111728.1 | - | 16 | 2069 | 40029118 | ||
BX647211.1 | - | 15 | 1731 | |||
CR749542.1 | - | 18 | 1979 |