Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERP44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015051.2 | NP_055866.1 | 8 | 896 | ||
GenBank mRNA | AB011145.1 | - | 8 | 820 | ||
AJ344330.2 | - | 8 | 807 | |||
AK075024.1 | - | 8 | 884 | |||
AY359048.1 | - | 8 | 820 | |||
BC005374.1 | - | 8 | 857 | 3934201 | ||
BX648519.1 | - | 9 | 987 |