Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other AMZ2 SIRNA Assays ›
Gene Symbol:
AMZ2
Gene Name:
archaelysin family metallopeptidase 2
Gene Aliases:
-
Chromosome Location:
Chr.17: 68205486 - 68257164 on Build GRCh38
UniGene ID:
Hs.293560
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
289774
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: AMZ2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001033569.1
NP_001028741.1
3
197
NM_001033570.1
NP_001028742.1
3
193
NM_001033571.1
NP_001028743.1
3
169
NM_001033572.1
NP_001028744.1
3
165
NM_001033574.1
NP_001028746.1
2
750
NM_001289054.1
NP_001275983.1
3
177
NM_001289056.1
NP_001275985.1
3
268
NM_016627.4
NP_057711.3
2
750
XM_005257432.4
XP_005257489.1
2
801
XM_005257433.3
XP_005257490.1
3
199
XM_005257435.4
XP_005257492.1
3
141
XM_005257436.3
XP_005257493.1
3
528
XM_006721940.2
XP_006722003.1
3
129
XM_017024723.1
XP_016880212.1
3
388
XM_017024724.1
XP_016880213.1
3
137
XM_017024725.1
XP_016880214.1
2
703
XM_017024726.1
XP_016880215.1
3
164
XM_017024727.1
XP_016880216.1
3
165
XM_017024728.1
XP_016880217.1
2
232
GenBank mRNA
AJ635358.1
-
1
48
AK090981.1
-
1
1036
AK126146.1
-
2
1180
BC050709.1
-
3
129
6060235
BC056271.1
-
3
162
4102397
BF978559.1
-
197
BM450564.1
-
3
138
BQ432895.1
-
1
37
CN278872.1
-
3
163
CN278874.1
-
3
165
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
proteolysis
metallopeptidase activity
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-4vtwv:80/10.239.167.181:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline