Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRCH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243963.1 | NP_001230892.1 | 20 | 4422 | ||
NM_020871.3 | NP_065922.3 | 21 | 4473 | |||
XM_006724724.3 | XP_006724787.2 | 21 | 4455 | |||
GenBank mRNA | AB040928.1 | - | 17 | 3655 | ||
AK022128.1 | - | 13 | 3230 | |||
BC042004.1 | - | 1 | 2210 | |||
BX647193.1 | - | 21 | 4473 |