Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF512B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020713.2 | NP_065764.1 | 3 | 270 | ||
XM_005260226.3 | XP_005260283.1 | 4 | 342 | |||
XM_011528929.2 | XP_011527231.1 | 3 | 232 | |||
XM_011528930.2 | XP_011527232.1 | 3 | 459 | |||
XM_011528933.2 | XP_011527235.1 | 3 | 455 | |||
GenBank mRNA | AB033022.1 | - | 1 | 89 | ||
BC125128.1 | - | 3 | 261 | 40119047 | ||
BC125129.1 | - | 3 | 260 | 40119053 |