Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF658 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317916.1 | NP_001304845.1 | 5 | 1446 | ||
NM_033160.6 | NP_149350.3 | 5 | 1289 | |||
NR_134255.1 | 4 | 1334 | ||||
XM_005272515.4 | XP_005272572.1 | 5 | 1396 | |||
XM_011545679.2 | XP_011543981.1 | 5 | 1220 | |||
XM_017014614.1 | XP_016870103.1 | 4 | 3582 | |||
GenBank mRNA | AK022636.1 | - | 1 | 136 | ||
AK024222.1 | - | 1 | 84 | |||
AK057375.1 | - | 5 | 1236 | |||
AK292150.1 | - | 5 | 1428 | |||
AL110217.1 | - | 1 | 59 | |||
BC031626.1 | - | 5 | 1370 | 5172270 | ||
BX479383.1 | - | 1 | 499 |