Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FMNL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052905.3 | NP_443137.2 | 26 | 4328 | ||
XM_005246263.2 | XP_005246320.1 | 27 | 4489 | |||
XM_005246265.3 | XP_005246322.1 | 27 | 4442 | |||
XM_011510530.1 | XP_011508832.1 | 27 | 4416 | |||
XM_011510531.1 | XP_011508833.1 | 28 | 4510 | |||
XM_011510532.2 | XP_011508834.1 | 28 | 4542 | |||
XM_011510533.2 | XP_011508835.1 | 28 | 4463 | |||
XM_011510534.1 | XP_011508836.1 | 26 | 4347 | |||
XM_011510535.2 | XP_011508837.1 | 26 | 4263 | |||
XM_011510536.2 | XP_011508838.1 | 21 | 3367 | |||
GenBank mRNA | AB067489.1 | - | 26 | 4099 | ||
AK094865.1 | - | 10 | 1883 | |||
AK131316.1 | - | 13 | 2620 | |||
AL834396.1 | - | 24 | 3712 | |||
BC014343.1 | - | 1 | 405 | |||
BC167159.1 | - | 26 | 4099 | 8862581 |