Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XIRP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199144.1 | NP_001186073.1 | 7 | 1064 | ||
NM_152381.5 | NP_689594.4 | 9 | 1466 | |||
XM_017003309.1 | XP_016858798.1 | 7 | 961 | |||
GenBank mRNA | AY820969.1 | - | 7 | 933 | ||
BC018673.1 | - | 4 | 520 | |||
EF119711.1 | - | 7 | 1064 |