Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAPLN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173462.3 | NP_775733.3 | 26 | 3983 | ||
XM_011537290.2 | XP_011535592.1 | 27 | 4064 | |||
XM_011537291.2 | XP_011535593.1 | 28 | 4111 | |||
XM_011537292.2 | XP_011535594.1 | 27 | 3987 | |||
XM_011537293.2 | XP_011535595.1 | 26 | 4030 | |||
XM_011537294.2 | XP_011535596.1 | 26 | 3988 | |||
XM_011537295.2 | XP_011535597.1 | 24 | 3746 | |||
XM_017021749.1 | XP_016877238.1 | 24 | 3793 | |||
XM_017021750.1 | XP_016877239.1 | 26 | 3986 | |||
GenBank mRNA | AK125658.1 | - | 6 | 1163 | ||
AK127354.1 | - | 12 | 2132 | |||
AK131073.1 | - | 19 | 3276 | |||
AK294560.1 | - | 26 | 4068 | |||
AY358330.1 | - | 10 | 1888 | |||
BC136623.1 | - | 26 | 3607 | 9020613 | ||
BC144315.1 | - | 23 | 3532 | |||
BX537757.1 | - | 12 | 2109 |