Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUBB2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178012.4 | NP_821080.1 | 2 | 328 | ||
GenBank mRNA | AK289433.1 | - | 2 | 249 | ||
AK308806.1 | - | 2 | 776 | |||
BC001352.2 | - | 2 | 328 | 2964590 | ||
BC063610.1 | - | 2 | 231 | 4941267 | ||
BT019930.1 | - | 2 | 137 | |||
CR456788.1 | - | 2 | 137 |