Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUBB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293212.1 | NP_001280141.1 | 4 | 1993 | ||
NM_001293213.1 | NP_001280142.1 | 5 | 1303 | |||
NM_001293214.1 | NP_001280143.1 | 3 | 1777 | |||
NM_001293215.1 | NP_001280144.1 | 4 | 1896 | |||
NM_001293216.1 | NP_001280145.1 | 4 | 1795 | |||
NM_178014.3 | NP_821133.1 | 4 | 1909 | |||
NR_120608.1 | 4 | 1310 | ||||
GenBank mRNA | AB062393.1 | - | 4 | 1737 | ||
AK098772.1 | - | 4 | 1754 | |||
AK297638.1 | - | 4 | 1377 | |||
AK316316.1 | - | 4 | 1791 | |||
BC001002.2 | - | 1 | 538 | |||
BC007605.1 | - | 4 | 1739 | 3350604 | ||
BC013374.2 | - | 4 | 1730 | 3946253 | ||
BC019924.1 | - | 4 | 1726 | 4998571 | ||
BC021909.1 | - | 4 | 1729 | 3509417 | ||
BC062532.1 | - | 4 | 1072 |