Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGBL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178563.3 | NP_848658.3 | 5 | 648 | ||
XM_011516122.2 | XP_011514424.1 | 4 | 454 | |||
XM_017012136.1 | XP_016867625.1 | 5 | 1970 | |||
XM_017012137.1 | XP_016867626.1 | 5 | 2252 | |||
XM_017012138.1 | XP_016867627.1 | 4 | 454 | |||
XM_017012139.1 | XP_016867628.1 | 4 | 454 | |||
XM_017012140.1 | XP_016867629.1 | 4 | 454 | |||
XM_017012141.1 | XP_016867630.1 | 4 | 1787 | |||
XM_017012142.1 | XP_016867631.1 | 4 | 454 | |||
XM_017012143.1 | XP_016867632.1 | 4 | 454 | |||
GenBank mRNA | AK302827.1 | - | 5 | 648 | ||
BC030651.2 | - | 5 | 623 | 5267615 |