Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF445 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181489.5 | NP_852466.1 | 8 | 1343 | ||
XM_005265102.2 | XP_005265159.1 | 7 | 1201 | |||
XM_005265103.3 | XP_005265160.1 | 5 | 473 | |||
XM_011533674.2 | XP_011531976.1 | 7 | 1166 | |||
XM_011533675.2 | XP_011531977.1 | 5 | 437 | |||
XM_017006345.1 | XP_016861834.1 | 7 | 1222 | |||
XM_017006346.1 | XP_016861835.1 | 7 | 1165 | |||
GenBank mRNA | AY262260.1 | - | 6 | 1146 | ||
AY295873.2 | - | 8 | 1343 | |||
BC101486.1 | - | 6 | 1057 | 8068992 | ||
BC143439.1 | - | 6 | 1022 | 9051947 | ||
L32162.1 | - | 7 | 1159 |