Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFSD4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181644.4 | NP_857595.3 | 10 | 3213 | ||
XM_011509229.2 | XP_011507531.1 | 9 | 2952 | |||
GenBank mRNA | AK090831.1 | - | 4 | 1940 | ||
AK127321.1 | - | 2 | 2849 | |||
AL713676.1 | - | 8 | 2519 | |||
AY358107.1 | - | 8 | 2599 | |||
BC016377.1 | - | 1 | 62 | |||
BC036549.2 | - | 10 | 3161 | 5264261 |