Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APOBEC3H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166002.2 | NP_001159474.2 | 5 | 755 | ||
NM_001166003.2 | NP_001159475.2 | 6 | 852 | |||
NM_181773.4 | NP_861438.3 | 5 | 758 | |||
XM_011529990.2 | XP_011528292.1 | 6 | 894 | |||
XM_011529991.2 | XP_011528293.1 | 6 | 891 | |||
XM_011529992.2 | XP_011528294.1 | 6 | 868 | |||
GenBank mRNA | BC069023.1 | - | 5 | 740 | 5441646 | |
BQ052182.1 | - | 5 | 697 | |||
BQ054330.1 | - | 6 | 798 | |||
CR456481.3 | - | 5 | 668 |