Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP26C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183374.2 | NP_899230.2 | 1 | 193 | ||
GenBank mRNA | AK131099.1 | - | 1 | 649 | ||
AY356349.1 | - | 1 | 193 |