Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LARP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018357.3 | NP_060827.2 | 1 | 352 | ||
NM_197958.2 | NP_932062.1 | 1 | 352 | |||
GenBank mRNA | AF443828.1 | - | 1 | 245 | ||
AF443829.1 | - | 1 | 245 | |||
AK002058.1 | - | 1 | 246 | |||
AL833876.1 | - | 1 | 206 | |||
BC006082.1 | - | 1 | 199 | 2961587 | ||
BC009446.1 | - | 1 | 199 | 3529516 | ||
BC014018.2 | - | 1 | 203 | 4127153 | ||
BC039153.1 | - | 1 | 208 | |||
BI913258.1 | - | 1 | 260 | |||
DN992782.1 | - | 1 | 357 |