Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEAD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003213.3 | NP_003204.2 | 7, 8 | 825 | ||
NM_201441.2 | NP_958849.1 | 6, 7 | 696 | |||
NM_201443.2 | NP_958851.1 | 5, 6 | 766 | |||
GenBank mRNA | AK222947.1 | - | 7, 8 | 712 | ||
AY101179.1 | - | 6, 7 | 678 | |||
BC015497.1 | - | 7, 8 | 723 | 3913870 | ||
BX640921.1 | - | 5, 6 | 432 | |||
KU178354.1 | - | 4, 5 | 314 | |||
U63824.1 | - | 7, 8 | 696 | |||
X94438.1 | - | 5, 6 | 512 |