Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF655 SIRNA Assays ›
Gene Symbol:
ZNF655
NUDCD3
Gene Name:
zinc finger protein 655
NudC domain containing 3
Gene Aliases:
VIK, VIK-1
NudCL
Chromosome Location:
Chr.7: 99557726 - 99576453 on Build GRCh38
Chromosome Location:
Chr.7: 44382366 - 44490875 on Build GRCh38
UniGene ID:
Hs.599798
Hs.488171
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID
290552
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF655
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001009958.1
NP_001009958.1
2
253
NM_001009960.1
NP_001009960.1
2
253
NM_001083956.1
NP_001077425.1
2
426
NM_001085366.1
NP_001078835.1
2
426
NM_001085367.1
NP_001078836.1
2
253
NM_001085368.1
NP_001078837.1
2
253
NM_024061.3
NP_076966.1
2
426
NM_138494.2
NP_612503.1
2
426
XM_017012598.1
XP_016868087.1
2
1113
XM_017012599.1
XP_016868088.1
2
271
XM_017012600.1
XP_016868089.1
2
284
XM_017012601.1
XP_016868090.1
2
270
XM_017012602.1
XP_016868091.1
2
1113
XM_017012603.1
XP_016868092.1
2
1346
XM_017012604.1
XP_016868093.1
2
263
XM_017012605.1
XP_016868094.1
2
147
XM_017012606.1
XP_016868095.1
2
284
XM_017012607.1
XP_016868096.1
2
270
XM_017012608.1
XP_016868097.1
2
1112
XM_017012609.1
XP_016868098.1
2
253
XM_017012610.1
XP_016868099.1
2
234
XM_017012611.1
XP_016868100.1
2
1122
XM_017012612.1
XP_016868101.1
2
1119
XM_017012613.1
XP_016868102.1
1
60
GenBank mRNA
AK027114.1
-
2
244
AK057245.1
-
3
152
AK225976.1
-
2
179
AK304785.1
-
2
130
AK314593.1
-
2
269
AK316024.1
-
2
244
AY099353.1
-
2
252
BC000823.1
-
2
196
BC004288.1
-
2
166
BC007378.2
-
2
120
BC037407.1
-
2
216
5752559
BC063478.1
-
2
426
BE293580.1
-
2
253
BI548533.1
-
2
173
BI912391.1
-
2
172
BM829333.1
-
2
280
BX490067.1
-
2
182
DN993232.1
-
2
189
Gene Symbol: NUDCD3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
GenBank mRNA
AK127163.1
-
126
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
negative regulation of G1/S transition of mitotic cell cycle
molecular_function
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-7q954:80/10.239.162.42:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0