Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC74B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258307.1 | NP_001245236.1 | 8 | 1092 | ||
NM_207310.2 | NP_997193.1 | 8 | 1290 | |||
XM_005263840.2 | XP_005263897.1 | 8 | 1315 | |||
XM_005263842.2 | XP_005263899.1 | 8 | 1117 | |||
XM_006712833.2 | XP_006712896.1 | 8 | 1327 | |||
XM_006712834.3 | XP_006712897.1 | 8 | 1267 | |||
XM_006712835.2 | XP_006712898.1 | 8 | 1129 | |||
XM_006712837.2 | XP_006712900.1 | 8 | 1069 | |||
XM_011512142.2 | XP_011510444.1 | 6 | 1558 | |||
GenBank mRNA | AK057609.1 | - | 8 | 1214 | ||
AK297246.1 | - | 7 | 2202 | |||
AL133619.1 | - | 4 | 3126 | |||
BC067771.1 | - | 8 | 1037 | 4824929 | ||
CD359923.1 | - | 3 | 289 |