Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LSM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021177.4 | NP_067000.1 | 5 | 463 | ||
GenBank mRNA | AF136977.1 | - | 5 | 448 | ||
AF182288.1 | - | 5 | 282 | |||
AF196468.1 | - | 5 | 428 | |||
AJ245416.1 | - | 5 | 454 | |||
BC009192.2 | - | 5 | 445 | 3842537 | ||
BP203102.1 | - | 5 | 463 | |||
BU189898.1 | - | 5 | 454 | |||
CR542146.1 | - | 4 | 236 | |||
CR542157.1 | - | 4 | 236 |