Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113182.2 | NP_001106653.1 | 11 | 3662 | ||
NM_001199455.1 | NP_001186384.1 | 11 | 3457 | |||
NM_001199456.1 | NP_001186385.1 | 10 | 2065 | |||
NM_001291986.1 | NP_001278915.1 | 12 | 3754 | |||
NM_005104.3 | NP_005095.1 | 11 | 3749 | |||
GenBank mRNA | AL832722.1 | - | 12 | 3689 | ||
BC063840.1 | - | 11 | 2322 | 6181728 | ||
BX647233.1 | - | 10 | 2065 | |||
BX648109.1 | - | 11 | 3663 | |||
D42040.1 | - | 11 | 3749 | |||
M80613.1 | - | 10 | 3084 | |||
X62083.1 | - | 10 | 3084 |