Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC4A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021196.3 | NP_067019.3 | 2 | 154 | ||
NM_133478.2 | NP_597812.1 | 7 | 550 | |||
GenBank mRNA | AF243499.2 | - | 2 | 152 | ||
AF293337.2 | - | 6 | 499 | |||
AF293338.2 | - | 2 | 152 | |||
AF452248.1 | - | 4 | 333 | |||
AF453528.1 | - | 4 | 342 |