Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSTC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267817.1 | NP_001254746.1 | 3 | 391 | ||
NM_001267818.1 | NP_001254747.1 | 5 | 663 | |||
NM_021227.3 | NP_067050.1 | 4 | 589 | |||
GenBank mRNA | AF201937.1 | - | 4 | 581 | ||
AF343342.1 | - | 4 | 518 | |||
BC016321.1 | - | 4 | 577 | 4075646 | ||
BC054857.1 | - | 4 | 579 | 4093030 | ||
BM471860.1 | - | 517 | ||||
BM541430.1 | - | 4 | 540 | |||
BM546028.1 | - | 4 | 579 | |||
CB110420.1 | - | 3 | 336 | |||
CN430159.1 | - | 5 | 602 |