Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF410 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242924.1 | NP_001229853.1 | 6 | 941 | ||
NM_001242926.1 | NP_001229855.1 | 5 | 890 | |||
NM_001242927.1 | NP_001229856.1 | 4 | 671 | |||
NM_001242928.1 | NP_001229857.1 | 5 | 890 | |||
NM_021188.2 | NP_067011.1 | 5 | 890 | |||
NR_040251.1 | 5 | 858 | ||||
GenBank mRNA | AK223261.1 | - | 5 | 646 | ||
AK293350.1 | - | 6 | 697 | |||
AK298307.1 | - | 6 | 613 | |||
AK298520.1 | - | 5 | 666 | |||
AK299138.1 | - | 4 | 669 | |||
AK301382.1 | - | 5 | 543 | |||
AK303834.1 | - | 5 | 556 | |||
AK309957.1 | - | 6 | 686 | |||
AK310031.1 | - | 4 | 432 | |||
AK315206.1 | - | 5 | 648 | |||
BC004357.1 | - | 5 | 634 | |||
BC050683.1 | - | 5 | 700 | 5752452 | ||
U90919.1 | - | 5 | 622 |