Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LSM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021177.4 | NP_067000.1 | 5 | 675 | ||
GenBank mRNA | AF136977.1 | - | 5 | 660 | ||
AF182288.1 | - | 5 | 493 | |||
AF196468.1 | - | 5 | 640 | |||
AJ245416.1 | - | 5 | 666 | |||
BC009192.2 | - | 5 | 657 | 3842537 |