Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC181 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300968.1 | NP_001287897.1 | 4 | 879 | ||
NM_001300969.1 | NP_001287898.1 | 3 | 581 | |||
NM_021179.2 | NP_067002.1 | 3 | 581 | |||
XM_005245383.1 | XP_005245440.1 | 4 | 879 | |||
XM_011509827.2 | XP_011508129.1 | 3 | 563 | |||
XM_017001938.1 | XP_016857427.1 | 5 | 2512 | |||
XM_017001939.1 | XP_016857428.1 | 3 | 544 | |||
XM_017001940.1 | XP_016857429.1 | 3 | 563 | |||
GenBank mRNA | AK098712.1 | - | 3 | 524 | ||
AK223354.1 | - | 3 | 524 | |||
AK313598.1 | - | 4 | 879 | |||
AL049687.1 | - | 3 | 539 | |||
BC026073.2 | - | 3 | 581 | 4827122 |