Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CADM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127173.1 | NP_001120645.1 | 9 | 1332 | ||
NM_021189.3 | NP_067012.1 | 10 | 1471 | |||
GenBank mRNA | AF062733.2 | - | 10 | 1428 | ||
AF363367.1 | - | 9 | 1330 | |||
AK001560.1 | - | 6 | 730 | |||
AK289541.1 | - | 10 | 1471 | |||
AY358332.1 | - | 9 | 1336 | |||
BC033819.1 | - | 9 | 1280 | 5199627 |