Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021201.4 | NP_067024.1 | 7 | 1255 | ||
NM_206938.1 | NP_996821.1 | 6 | 1120 | |||
NM_206939.1 | NP_996822.1 | 7 | 1207 | |||
NM_206940.1 | NP_996823.1 | 6 | 1072 | |||
GenBank mRNA | AF237916.1 | - | 7 | 1179 | ||
AK075106.1 | - | 7 | 1259 | |||
AK297027.1 | - | 6 | 1056 |