Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00474 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024032.1 | 3 | 461 | |||
GenBank mRNA | AB021923.1 | - | 3 | 571 | ||
BC104240.1 | - | 3 | 461 | |||
BC104241.1 | - | 3 | 460 | |||
BC112996.1 | - | 3 | 461 | |||
BC112997.1 | - | 3 | 461 | |||
BC128039.1 | - | 3 | 461 |