Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRUNE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303229.1 | NP_001290158.1 | 4 | 707 | ||
NM_001303242.1 | NP_001290171.1 | 5 | 910 | |||
NM_021222.2 | NP_067045.1 | 5 | 910 | |||
NR_130130.1 | 2 | 429 | ||||
NR_130135.1 | 3 | 522 | ||||
XM_005245393.4 | XP_005245450.1 | 5 | 798 | |||
XM_011509832.2 | XP_011508134.1 | 3 | 599 | |||
XM_017001955.1 | XP_016857444.1 | 5 | 798 | |||
XM_017001956.1 | XP_016857445.1 | 3 | 508 | |||
GenBank mRNA | AF123538.1 | - | 2 | 343 | ||
AF123539.1 | - | 4 | 591 | |||
AK027875.1 | - | 2 | 202 | |||
AK298273.1 | - | 1 | 38 | |||
AK315120.1 | - | 5 | 841 | |||
BC014886.1 | - | 4 | 615 | 3907577 | ||
BC025304.1 | - | 5 | 804 | 5458046 | ||
DA942287.1 | - | 3 | 370 |