Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC5A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001305005.2 | NP_001291934.1 | 3 | 499 | ||
NM_001305006.2 | NP_001291935.1 | 2 | 273 | |||
NM_001305007.2 | NP_001291936.1 | 3 | 502 | |||
NM_021815.4 | NP_068587.1 | 3 | 502 | |||
XM_017004629.1 | XP_016860118.1 | 1 | 3766 | |||
GenBank mRNA | AB043997.1 | - | 3 | 477 | ||
AF276871.1 | - | 2 | 201 | |||
AJ401466.1 | - | 2 | 219 | |||
AK300801.1 | - | 2 | 263 | |||
AK315280.1 | - | 3 | 491 | |||
BC111524.1 | - | 3 | 444 | 40034612 | ||
BC111525.1 | - | 3 | 449 | 40034618 | ||
DA714970.1 | - | 3 | 503 |