Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPBP1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021639.4 | NP_067652.1 | 5 | 1481 | ||
XM_017002019.1 | XP_016857508.1 | 5 | 1542 | |||
GenBank mRNA | AF177334.1 | - | 3 | 299 | ||
AK024972.1 | - | 3 | 288 | |||
AK225197.1 | - | 3 | 288 | |||
AL834194.1 | - | 3 | 1131 | |||
BC015713.1 | - | 3 | 380 | 3909296 | ||
BC022218.1 | - | 3 | 564 | 3900818 | ||
BC052257.1 | - | 2 | 129 | 6297561 | ||
BC068578.1 | - | 5 | 1461 |