Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FAM3A SIRNA Assays ›
Gene Symbol:
FAM3A
Gene Name:
family with sequence similarity 3 member A
Gene Aliases:
2.19, DLD, DXS560S, XAP-7
Chromosome Location:
Chr.X: 154506159 - 154516249 on Build GRCh38
UniGene ID:
Hs.289108
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
29441
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FAM3A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001171132.2
NP_001164603.1
5, 6
648
NM_001171133.2
NP_001164604.1
4, 5
534
NM_001171134.2
NP_001164605.1
4, 5
745
NM_001282311.1
NP_001269240.1
5, 6
843
NM_001282312.1
NP_001269241.1
5, 6
589
NM_021806.3
NP_068578.2
4, 5
745
XM_005274714.3
XP_005274771.1
6, 7
693
XM_005274716.3
XP_005274773.1
6, 7
693
XM_005277879.4
XP_005277936.1
5, 6
654
XM_006724831.2
XP_006724894.1
6, 7
693
XM_006724832.3
XP_006724895.1
5, 6
654
XM_006724833.2
XP_006724896.1
6, 7
693
XM_006724834.2
XP_006724897.1
5, 6
806
XM_011531185.2
XP_011529487.1
3, 4
251
XM_017029699.1
XP_016885188.1
4, 5
966
XM_017029700.1
XP_016885189.1
4, 5
965
XM_017029701.1
XP_016885190.1
5, 6
868
XM_017029702.1
XP_016885191.1
4, 5
481
GenBank mRNA
AK024601.1
-
5, 6
605
AK057945.1
-
5, 6
463
AK124890.1
-
1, 2
2936
AK294116.1
-
4, 5
708
AK314682.1
-
4, 5
663
AL539139.3
-
5, 6
840
BC002934.1
-
5, 6
747
BC008912.2
-
4, 5
396
3639043
BM457676.1
-
5, 6
630
CX867071.1
-
4, 5
452
X87193.1
-
4, 5
552
Back To Top
More Information
Panther Classification:
Molecular Function -
antimicrobial response protein
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
molecular_function
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-7q954:80/10.239.162.42:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0