Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPCS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021928.3 | NP_068747.1 | 4, 5 | 557 | ||
GenBank mRNA | AK026302.1 | - | 4, 5 | 532 | ||
AK092634.1 | - | 3, 4 | 1540 | |||
AL136660.1 | - | 4, 5 | 527 | |||
AY359044.1 | - | 4, 5 | 515 | |||
BC047290.1 | - | 4, 5 | 500 | 5224927 | ||
BC053883.1 | - | 3, 4 | 2451 |