Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPBP1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021639.4 | NP_067652.1 | 12 | 2563 | ||
XM_017002019.1 | XP_016857508.1 | 11 | 2465 | |||
GenBank mRNA | AF177334.1 | - | 10 | 1381 | ||
AK024972.1 | - | 10 | 1370 | |||
AK027212.1 | - | 4 | 1946 | |||
AK225197.1 | - | 10 | 1370 | |||
AL834194.1 | - | 10 | 2213 | |||
BC015713.1 | - | 10 | 1462 | 3909296 | ||
BC022218.1 | - | 10 | 1646 | 3900818 | ||
BC052257.1 | - | 9 | 1211 | 6297561 | ||
BC068578.1 | - | 12 | 2543 |