Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021927.2 | NP_068746.2 | 2, 3 | 492 | ||
XM_011513732.2 | XP_011512034.1 | 2, 3 | 513 | |||
GenBank mRNA | AK023282.1 | - | 2, 3 | 442 | ||
AK225314.1 | - | 2, 3 | 447 | |||
BC036768.1 | - | 2, 3 | 453 | 5744283 | ||
DA997676.1 | - | 2, 3 | 492 |