Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: LAMTOR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243736.1 | NP_001230665.1 | 5 | 414 | ||
NM_021970.3 | NP_068805.1 | 5 | 435 | |||
NR_024170.1 | 4 | 389 | ||||
GenBank mRNA | AF201947.1 | - | 5 | 345 | ||
AK223311.1 | - | 5 | 326 | |||
AK311755.1 | - | 5 | 296 | |||
AL544981.3 | - | 5 | 244 | |||
BC026245.1 | - | 5 | 328 | 4808855 | ||
BC110402.1 | - | 4 | 255 | |||
BI562630.1 | - | 4 | 392 |