Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GBGT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282629.1 | NP_001269558.1 | 7 | 1113 | ||
NM_001282632.1 | NP_001269561.1 | 6 | 1081 | |||
NM_001288572.1 | NP_001275501.1 | 7 | 1169 | |||
NM_001288573.1 | NP_001275502.1 | 7 | 1169 | |||
NM_021996.5 | NP_068836.2 | 7 | 1132 | |||
GenBank mRNA | AK074639.1 | - | 7 | 1080 | ||
AK074944.1 | - | 5 | 2095 | |||
AK291498.1 | - | 7 | 1101 | |||
AK303825.1 | - | 6 | 1050 | |||
AK307687.1 | - | 7 | 1138 | |||
AK314097.1 | - | 7 | 1101 | |||
AY358175.1 | - | 7 | 1081 | |||
BC032499.1 | - | 7 | 1084 | 5223113 |