Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VIPAS39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193314.1 | NP_001180243.1 | 7, 8 | 1052 | ||
NM_001193315.1 | NP_001180244.1 | 7, 8 | 652 | |||
NM_001193316.1 | NP_001180245.1 | 6, 7 | 505 | |||
NM_001193317.1 | NP_001180246.1 | 7, 8 | 829 | |||
NM_022067.3 | NP_071350.2 | 8, 9 | 923 | |||
XM_017021580.1 | XP_016877069.1 | 7, 8 | 643 | |||
XM_017021581.1 | XP_016877070.1 | 7, 8 | 777 | |||
GenBank mRNA | AK022769.1 | - | 7, 8 | 600 | ||
AK022925.1 | - | 7, 8 | 578 | |||
AK098254.1 | - | 7, 8 | 1013 | |||
AK222706.1 | - | 7, 8 | 598 | |||
AK297388.1 | - | 7, 8 | 754 | |||
AK298354.1 | - | 6, 7 | 451 | |||
BC015054.1 | - | 8, 9 | 844 | 3920459 | ||
CR457294.1 | - | 6, 7 | 506 |