Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PCIF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022104.3 | NP_071387.1 | 8 | 1045 | ||
XM_011528980.2 | XP_011527282.1 | 8 | 1096 | |||
XM_011528981.2 | XP_011527283.1 | 8 | 1007 | |||
XM_017028013.1 | XP_016883502.1 | 8 | 1206 | |||
XM_017028014.1 | XP_016883503.1 | 8 | 1009 | |||
GenBank mRNA | AB050014.1 | - | 8 | 1025 | ||
AK056553.1 | - | 1023 | ||||
AK299435.1 | - | 8 | 1030 | |||
BC010005.2 | - | 8 | 1017 | 4131938 | ||
BC013365.2 | - | 8 | 1016 | 3640928 |