Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH2D4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174159.1 | NP_001167630.1 | 10 | 2368 | ||
NM_001174160.1 | NP_001167631.1 | 9 | 2133 | |||
NM_022071.3 | NP_071354.2 | 10 | 2518 | |||
XM_011544610.1 | XP_011542912.1 | 9 | 2564 | |||
XM_017013729.1 | XP_016869218.1 | 9 | 2286 | |||
GenBank mRNA | AK024620.1 | - | 8 | 1787 | ||
AK024799.1 | - | 10 | 2396 | |||
BC067117.1 | - | 7 | 1727 |